Merck
  • Synthesis and nicotinic acetylcholine receptor binding properties of bridged and fused ring analogues of epibatidine.
Synthesis and nicotinic acetylcholine receptor binding properties of bridged and fused ring analogues of epibatidine.
Journal of medicinal chemistry (2007-11-13)
F Ivy Carroll, T Philip Robinson, Lawrence E Brieaddy, Robert N Atkinson, S Wayne Mascarella, M Imad Damaj, Billy R Martin, Hernán A Navarro
ZUSAMMENFASSUNG
Epibatidine analogues 3- 5, possessing the pyridine ring fused to the 2,3 position of the 7-azabicyclo[2.2.1]heptane ring, and analogue 8a, possessing a benzene ring fused to the 5,6 position, were synthesized by procedures involving key steps of trapping 2,3-pyridyne, 3,4-pyridyne, and benzyne with tert-butyl 1 H-pyrrole-1-carboxylate. Two epibatidine analogues, 6 and 7, which have the 2'-chloropyridine ring bridged to the 7-azabicyclo[2.2.1]heptane ring via a methylene group, were synthesized, where the key step was an intramolecular reductive palladium-catalyzed Heck-type coupling. Even though the conformationally restricted epibatidine analogues, 3- 7, and the benzo analogue 8a possess nAChR pharmacophore features thought to be needed for alpha(4)beta(2) binding, they all showed low affinity for nAChRs relative to epibatidine. These studies provide new information concerning the pharmacophore for nAChRs and suggest that nitrogen lone-pair directionality and steric factors may be important. Interestingly, N-methylepibatidine, prepared as a standard compound for the study of bridged analogues 6 and 7, was a potent nAChR mixed agonist antagonist.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Nikotin, ≥99% (GC), liquid
Supelco
(−)-Nikotin, PESTANAL®, analytical standard
Supelco
(−)-Nikotin -Lösung, 1.0 mg/mL, analytical standard, for drug analysis