- Intermediate analogue inhibitors of mandelate racemase: N-Hydroxyformanilide and cupferron.
Intermediate analogue inhibitors of mandelate racemase: N-Hydroxyformanilide and cupferron.
Bioorganic & medicinal chemistry letters (2006-10-24)
Jennifer R Bourque, Rodney K M Burley, Stephen L Bearne
PMID17055725
ZUSAMMENFASSUNG
Mandelate racemase (MR) catalyzes the 1,1-proton transfer that interconverts the enantiomers of mandelate. The transition state/intermediate analogues N-hydroxyformanilide (K(i)=2.79+/-0.19 microM) and cupferron (K(i)=2.67+/-0.09 microM) are identified as potent competitive inhibitors of MR. The pH-pK(i) profile indicates that MR can bind either the protonated or deprotonated forms of N-hydroxyformanilide, with a 10-fold greater affinity for the latter form.