Merck
  • Carbonic anhydrase activators: an activation study of the human mitochondrial isoforms VA and VB with amino acids and amines.
Carbonic anhydrase activators: an activation study of the human mitochondrial isoforms VA and VB with amino acids and amines.
Bioorganic & medicinal chemistry letters (2006-12-19)
Daniela Vullo, Isao Nishimori, Alessio Innocenti, Andrea Scozzafava, Claudiu T Supuran
ZUSAMMENFASSUNG
The mitochondrial isozymes of human carbonic anhydrase (hCA, EC 4.2.1.1), hCA VA and hCA VB, were investigated for activation with a series of amino acids and amines. D-His, L-DOPA, histamine, dopamine, and 4-(2-aminoethyl)morpholine were excellent hCA VA activators, with KAs in the range of 10-130 nM. Good hCA VB activating effects were identified for L-His, D-Phe, D-DOPA, L-Trp, L-Tyr, serotonin, and 2-(2-aminoethyl)-pyridine, with KAs in the range of 44-110 nM. All these activators enhanced kcat, having no effect on KM, favoring thus the rate-determining step in the catalytic cycle, the proton transfer reactions between the active site and environment. The activation pattern of the two mitochondrial isoforms is very different from each other and as compared to those of the cytosolic isoforms hCA I and II.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Histidin, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
L-Tyrosin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Histidin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidin, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Phenylalanin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
SAFC
L-Histidin
Sigma-Aldrich
Histamin, ≥97.0%
Sigma-Aldrich
D-Phenylalanin, ≥98% (HPLC)
Sigma-Aldrich
L-Phenylalanin, reagent grade, ≥98%
Sigma-Aldrich
2-Picolylamin, 99%
Sigma-Aldrich
1-(2-Aminoethyl)piperazin, 99%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Phenylalanin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
4-(2-Aminoethyl)-morpholin, 99%
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
SAFC
L-Tyrosin
Sigma-Aldrich
3,4-Dihydroxy-D-Phenylalanin, powder, ≥95%
Sigma-Aldrich
D-Histidin, ≥98% (TLC)
Sigma-Aldrich
2-(2-Pyridyl)ethylamin, 95%
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosin, FG
Sigma-Aldrich
L-Phenylalanin, 99%, FCC
Supelco
L-Phenylalanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland