Merck
  • Ataxia-telangiectasia: chronic activation of damage-responsive functions is reduced by alpha-lipoic acid.
Ataxia-telangiectasia: chronic activation of damage-responsive functions is reduced by alpha-lipoic acid.
Oncogene (2001-04-21)
M Gatei, D Shkedy, K K Khanna, T Uziel, Y Shiloh, T K Pandita, M F Lavin, G Rotman
ZUSAMMENFASSUNG
Cells from patients with the genetic disorder ataxia-telangiectasia (A-T) are hypersensitive to ionizing radiation and radiomimetic agents, both of which generate reactive oxygen species capable of causing oxidative damage to DNA and other macromolecules. We describe in A-T cells constitutive activation of pathways that normally respond to genotoxic stress. Basal levels of p53 and p21(WAF1/CIP1), phosphorylation on serine 15 of p53, and the Tyr15-phosphorylated form of cdc2 are chronically elevated in these cells. Treatment of A-T cells with the antioxidant alpha-lipoic acid significantly reduced the levels of these proteins, pointing to the involvement of reactive oxygen species in their chronic activation. These findings suggest that the absence of functional ATM results in a mild but continuous state of oxidative stress, which could account for several features of the pleiotropic phenotype of A-T.