Merck
  • CMT-3 targets different α-synuclein aggregates mitigating their toxic and inflammogenic effects.
CMT-3 targets different α-synuclein aggregates mitigating their toxic and inflammogenic effects.
Scientific reports (2020-11-22)
Florencia González-Lizárraga, Diego Ploper, César L Ávila, Sergio B Socías, Mauricio Dos-Santos-Pereira, Belén Machín, Elaine Del-Bel, Patrick Pierre Michel, Lía I Pietrasanta, Rita Raisman-Vozari, Rosana Chehín
ZUSAMMENFASSUNG
Parkinson's disease (PD) is a neurodegenerative disorder for which only symptomatic treatments are available. Repurposing drugs that target α-synuclein aggregation, considered one of the main drivers of PD progression, could accelerate the development of disease-modifying therapies. In this work, we focused on chemically modified tetracycline 3 (CMT-3), a derivative with reduced antibiotic activity that crosses the blood-brain barrier and is pharmacologically safe. We found that CMT-3 inhibited α-synuclein amyloid aggregation and led to the formation of non-toxic molecular species, unlike minocycline. Furthermore, CMT-3 disassembled preformed α-synuclein amyloid fibrils into smaller fragments that were unable to seed in subsequent aggregation reactions. Most interestingly, disaggregated species were non-toxic and less inflammogenic on brain microglial cells. Finally, we modelled the interactions between CMT-3 and α-synuclein aggregates by molecular simulations. In this way, we propose a mechanism for fibril disassembly. Our results place CMT-3 as a potential disease modifier for PD and possibly other synucleinopathies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Chlorameisensäure-2,2,2-trichlorethylester, 98%