Merck
  • Functional single-nucleotide polymorphism of epidermal growth factor is associated with the development of Barrett's esophagus and esophageal adenocarcinoma.
Functional single-nucleotide polymorphism of epidermal growth factor is associated with the development of Barrett's esophagus and esophageal adenocarcinoma.
Journal of human genetics (2011-12-02)
Vivianda Menke, Raymond G J Pot, Leon M G Moons, Katinka P M van Zoest, Bettina Hansen, Herman van Dekken, Peter D Siersema, Johannes G Kusters, Ernst J Kuipers
ZUSAMMENFASSUNG
Reflux esophagitis (RO) and Barrett's esophagus (BO) can cause esophageal adenocarcinoma (OAC). The esophageal mucosa in the RO-BO-OAC cascade is chronically exposed to gastro-esophageal reflux. Epidermal growth factor (EGF) has an important role in the protection and repair of mucosal damage, and non-physiologic levels are associated with gastrointestinal tumors. The aim is to determine the functional effect of EGF gene polymorphisms on RO, BO and OAC development. A cohort of 871 unrelated Dutch Caucasians consisted of 198 healthy controls, 298 RO patients, 246 BO patients and 129 OAC patients. The frequency of the EGF-production-associated 5'UTR A+61G polymorphism was determined in these four groups. EGF immunohistochemistry was performed on BO biopsies. EGF expression was significantly lower in the G/G genotype compared with the A/G (P=0.008) and A/A (P=0.002) group. The G/G genotype was significantly more prevalent in RO (odds ratios (OR)=2.6; 95% confidence intervals (95% CI): 1.3-5.2), BO (OR=3.0; 95% CI: 1.5-6.2) and OAC (OR=4.1; 95% CI: 1.8-9.7) than in controls. The G allele is associated with reduced EGF expression and increased risk for RO, BO and OAC development. This indicates that reduced mucosal protection resulting from genetically decreased EGF expression enhances esophageal tumor development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Epidermal Growth Factor antibody, Mouse monoclonal, clone EGF-10, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid