- Lipopeptide Epimers and a Phthalide Glycerol Ether with AChE Inhibitory Activities from the Marine-Derived Fungus Cochliobolus Lunatus SCSIO41401.
Lipopeptide Epimers and a Phthalide Glycerol Ether with AChE Inhibitory Activities from the Marine-Derived Fungus Cochliobolus Lunatus SCSIO41401.
Marine drugs (2020-11-05)
Yu Dai, Kunlong Li, Jianglian She, Yanbo Zeng, Hao Wang, Shengrong Liao, Xiuping Lin, Bin Yang, Junfeng Wang, Huaming Tao, Haofu Dai, Xuefeng Zhou, Yonghong Liu
PMID33143384
ZUSAMMENFASSUNG
A pair of novel lipopeptide epimers, sinulariapeptides A (1) and B (2), and a new phthalide glycerol ether (3) were isolated from the marine algal-associated fungus Cochliobolus lunatus SCSIO41401, together with three known chromanone derivates (4-6). The structures of the new compounds, including the absolute configurations, were determined by comprehensive spectroscopic methods, experimental and calculated electronic circular dichroism (ECD), and Mo2 (OAc)4-induced ECD methods. The new compounds 1-3 showed moderate inhibitory activity against acetylcholinesterase (AChE), with IC50 values of 1.3-2.5 μM, and an in silico molecular docking study was also performed.