Merck
  • Synthesis and biological evaluation of new N-(2-hydroxy-4(or 5)-nitro/aminophenyl)benzamides and phenylacetamides as antimicrobial agents.
Synthesis and biological evaluation of new N-(2-hydroxy-4(or 5)-nitro/aminophenyl)benzamides and phenylacetamides as antimicrobial agents.
Bioorganic & medicinal chemistry (2007-01-16)
Tugba Ertan, Ilkay Yildiz, Semiha Ozkan, Ozlem Temiz-Arpaci, Fatma Kaynak, Ismail Yalcin, Esin Aki-Sener, Ufuk Abbasoglu
ZUSAMMENFASSUNG
A new series of N-(2-hydroxy-4(or 5)-nitro/aminophenyl)benzamide and phenylacetamide derivatives (1a-1n, 2a-2n) were synthesized and evaluated for antibacterial and antifungal activities against Staphylococcus aureus, Bacillus subtilis, Klebsiella pneumoniae, Pseudomonas aeruginosa, Escherichia coli, Candida albicans, and their drug-resistant isolate. Microbiological results indicated that the compounds possessed a broad spectrum of activity against the tested microorganisms at MIC values between 500 and 1.95 microg/ml. Benzamide derivative 1d exhibited the greatest activity with MIC values of 1.95, 3.9, and 7.8 microg/ml against drug-resistant B. subtilis, B. subtilis, and S. aureus, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin Trihydrat, 900-1050 μg/mg anhydrous basis (HPLC)
Supelco
Ampicillin Trihydrat, VETRANAL®, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard