- Outbreak of Rocky Mountain spotted fever in Córdoba, Colombia.
Outbreak of Rocky Mountain spotted fever in Córdoba, Colombia.
Memorias do Instituto Oswaldo Cruz (2011-02-23)
Marylin Hidalgo, Jorge Miranda, Damaris Heredia, Pilar Zambrano, Juan Fernando Vesga, Diana Lizarazo, Salim Mattar, Gustavo Valbuena
PMID21340366
ZUSAMMENFASSUNG
Rocky Mountain spotted fever (RMSF) is a tick-borne disease caused by the obligate intracellular bacterium Rickettsia rickettsii. Although RMSF was first reported in Colombia in 1937, it remains a neglected disease. Herein, we describe the investigation of a large cluster of cases of spotted fever rickettsiosis in a new area of Colombia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Adenovirus-Coating-Antikörper (Mischung, Klone 2/6 und 20/11), ascites fluid, Chemicon®, from mouse