Merck
Seroprevalence of anti-SARS-CoV-2 IgG antibodies in Kenyan blood donors.
Science (New York, N.Y.) (2020-11-13)
Sophie Uyoga, Ifedayo M O Adetifa, Henry K Karanja, James Nyagwange, James Tuju, Perpetual Wanjiku, Rashid Aman, Mercy Mwangangi, Patrick Amoth, Kadondi Kasera, Wangari Ng'ang'a, Charles Rombo, Christine Yegon, Khamisi Kithi, Elizabeth Odhiambo, Thomas Rotich, Irene Orgut, Sammy Kihara, Mark Otiende, Christian Bottomley, Zonia N Mupe, Eunice W Kagucia, Katherine E Gallagher, Anthony Etyang, Shirine Voller, John N Gitonga, Daisy Mugo, Charles N Agoti, Edward Otieno, Leonard Ndwiga, Teresa Lambe, Daniel Wright, Edwine Barasa, Benjamin Tsofa, Philip Bejon, Lynette I Ochola-Oyier, Ambrose Agweyu, J Anthony G Scott, George M Warimwe
ZUSAMMENFASSUNG
The spread of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) in Africa is poorly described. The first case of SARS-CoV-2 in Kenya was reported on 12 March 2020, and an overwhelming number of cases and deaths were expected, but by 31 July 2020, there were only 20,636 cases and 341 deaths. However, the extent of SARS-CoV-2 exposure in the community remains unknown. We determined the prevalence of anti-SARS-CoV-2 immunoglobulin G among blood donors in Kenya in April-June 2020. Crude seroprevalence was 5.6% (174 of 3098). Population-weighted, test-performance-adjusted national seroprevalence was 4.3% (95% confidence interval, 2.9 to 5.8%) and was highest in urban counties Mombasa (8.0%), Nairobi (7.3%), and Kisumu (5.5%). SARS-CoV-2 exposure is more extensive than indicated by case-based surveillance, and these results will help guide the pandemic response in Kenya and across Africa.