Merck
Novel cationic lipophilic peptides for oligodeoxynucleotide delivery.
Bioorganic & medicinal chemistry (2007-04-11)
Enoch Chan, Michael Amon, Robert J Marano, Norbert Wimmer, Philip S Kearns, Nicholas Manolios, P Elizabeth Rakoczy, Istvan Toth
ZUSAMMENFASSUNG
In search of new oligodeoxynucleotide (ODN) delivery agents, we evaluated novel peptides derived from core peptide H-GLRILLLKV-OH (CP). CP is a fragment designed from the T-cell antigen receptor (TCR) alpha-chain transmembrane sequence. CP was able to enter cells including T-cells and inhibited interleukin-2 (IL-2) production. To examine the effect of increased lipophilicity on cellular uptake and activity of CP, a lipoamino acid (2-aminododecanoic acid) was incorporated into peptide CP resulting in 2-aminodecanoyl-CP (LP). The toxicity of CP and LP was assessed by measuring the haemolytic activity. Neither compound caused any haemolysis of red blood cells. We have also compared the biological activities of the CP and LP. Using a T-cell antigen presentation assay, the more lipophilic LP caused greater inhibition of IL-2 production than the parent CP in the antigen stimulated T-cells. The LP also showed increased permeability than CP in the Caco-2 cell assay. We utilised the enhanced cell permeability property of LP in oligodeoxynucleotide ODN1 delivery. Isothermal titration calorimetry (ITC) suggested that CP and LP complex with ODN1 in a 12:1 (CP:ODN1) and 15:1 (LP:ODN1) ratio. These complexes were then transfected into human retinal pigment epithelial cells. The level of transfection was measured by the decreased production of the protein human vascular endothelial growth factor (hVEGF). The results revealed greater transfection efficiency for both CP and LP (47%, 55% more inhibition) compared to commercially available transfection agent cytofectin GSV. These results suggested that the CP and particularly its lipophilic analogue LP have the potential to be used as oligodeoxynucleotide delivery systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri