- Synthesis and structure-activity relationships of 16-modified analogs of 2-methoxyestradiol.
Synthesis and structure-activity relationships of 16-modified analogs of 2-methoxyestradiol.
Bioorganic & medicinal chemistry (2007-10-04)
Gregory E Agoston, Jamshed H Shah, Theresa M Lavallee, Xiaoguo Zhan, Victor S Pribluda, Anthony M Treston
PMID17910916
ZUSAMMENFASSUNG
A series of 16-modified 2-methoxyestradiol analogs were synthesized and evaluated for antiproliferative activity toward HUVEC and MDA-MB-231 cells, and for susceptibility to conjugation. In addition, the estrogenicity of these analogs was accessed by measuring cell proliferation of the estrogen-dependent cell line MCF7 in response to compound treatment. It was observed that antiproliferative activity dropped as the size of the 16 substituent increased. Selected analogs tested in glucuronidation assays had similar rates of clearance to 2-methoxyestradiol, but had enhanced clearance in sulfonate conjugation assays.
MATERIALIEN