- New GABA-modulating 1,2,4-oxadiazole derivatives and their anticonvulsant activity.
New GABA-modulating 1,2,4-oxadiazole derivatives and their anticonvulsant activity.
European journal of medicinal chemistry (2007-02-17)
Hans-Joachim Lankau, Klaus Unverferth, Christian Grunwald, Helge Hartenhauer, Kristina Heinecke, Katrin Bernöster, Rita Dost, Ute Egerland, Chris Rundfeldt
PMID17303289
ZUSAMMENFASSUNG
A series of 3- and 5-aryl-1,2,4-oxadiazole derivatives were prepared and tested for anticonvulsant activity in a variety of models. These 1,2,4-oxadiazoles exhibit considerable activity in both pentylenetetrazole (PTZ) and maximal electroshock seizure (MES) models. Compound 10 was protective in the PTZ model in rats with an oral ED(50) of 25.5mg/kg and in the MES model in rats with an oral ED(50) of 14.6mg/kg. Neurotoxicity (rotarod) was observed with an ED(50) of 335mg/kg. We found several oxadiazoles that acted as selective GABA potentiating compounds with no interaction to the benzodiazepine binding site.