Merck
Post-translational modifications in collagen type I of bone in a mouse model of aging.
Bone (2020-11-22)
Amy Creecy, Kyle L Brown, Kristie L Rose, Paul Voziyan, Jeffry S Nyman, Amy Creecy, Kyle L Brown, Kristie L Rose, Paul Voziyan, Jeffry S Nyman
ZUSAMMENFASSUNG
The fracture resistance of cortical bone and matrix hydration are known to decline with advanced aging. However, the underlying mechanisms remain poorly understood, and so we investigated levels of matrix proteins and post-translational modifications (PTM) of collagen I in extracts from the tibia of 6-mo. and 20-mo. old BALB/c mice (female and male analysis done separately). Liquid chromatography-tandem mass spectrometry (LC-MS/MS) analysis revealed that the levels of collagen I deamidation at specific asparagine (Asn) and glutamine (Gln) residues significantly increased with age. Other non-enzymatic PTMs such as carboxymethylation of lysine (CML) were detected as well, but the relative abundance did not vary with age. No significant age-related differences in the abundance of hydroxylysine glycosylation sites were found, but hydroxylation levels at a few of the numerous lysine and proline hydroxylation sites significantly changed by a small amount with age. We performed molecular modeling and dynamics (MD) simulations for three triple helical fragments representing collagen I regions with prominent age-dependent increases in deamidation as identified by LC-MS/MS of male extracts. These 3 fragments included deamidated Asn and Gln residues as follows: 1) an Asn428 site of the α2(I) chain in which deamidation levels increased from 4.4% at 6-mo. to 8.1% at 20-mo., 2) an Asn983 site of the α2(I) chain with a deamidation increase from 18.3% to 36.8% with age and an Asn1052 site of the α1(I) chain with consistent deamidation levels of ~60% across the age groups, and 3) a Gln410 site of the α1(I) chain that went from no detectable deamidation at 6-mo. to 2.7% at 20-mo. and a neighboring Asn421 site of the same chain with an age-related deamidation increase from 3.6% to 16.3%. The deamidation levels at these sites inversely correlated with an estimate of toughness determined from three-point bending tests of the femur mid-diaphysis. MD revealed that the sidechains become more negatively charged at deamidated sites and that deamidation alters hydrogen bonding with water along the collagen backbone while increasing water interactions with the aspartic and glutamic acid sidechains. Our findings suggest a new mechanism of the age-dependent reduction in the fracture resistance of cortical bone whereby deamidation of Asn and Glu residues redistributes bound water within collagen I triple helix.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Brij® L23 -Lösung, 30 % (w/v) in H2O
Sigma-Aldrich
Angiotensin I human, ≥90% (HPLC), powder, suitable for mammalian cell culture
Sigma-Aldrich
ProteoMass Angiotensin II MALDI-MS-Standard, vial of 10 nmol