Merck
  • A Feedforward Circuit Regulates Action Selection of Pre-mating Courtship Behavior in Female Drosophila.
A Feedforward Circuit Regulates Action Selection of Pre-mating Courtship Behavior in Female Drosophila.
Current biology : CB (2020-01-07)
Hiroshi Ishimoto, Azusa Kamikouchi
ZUSAMMENFASSUNG
In the early phase of courtship, female fruit flies exhibit an acute rejection response to avoid unfavorable mating. This pre-mating rejection response is evolutionarily paralleled across species, but the molecular and neuronal basis of that behavior is unclear. Here, we show that a putative incoherent feedforward circuit comprising ellipsoid body neurons, cholinergic R4d, and its repressor GABAergic R2/R4m neurons regulates the pre-mating rejection response in the virgin female Drosophila melanogaster. Both R4d and R2/R4m are positively regulated, via specific dopamine receptors, by a subset of neurons in the dopaminergic PPM3 cluster. Genetic deprivation of GABAergic signal via GABAA receptor RNA interference in this circuit induces a massive rejection response, whereas activation of GABAergic R2/R4m or suppression of cholinergic R4d increases receptivity. Moreover, glutamatergic signaling via N-methyl-d-aspartate receptors induces NO-mediated retrograde regulation potentially from R4d to R2/R4m, likely providing flexible control of the behavioral switching from rejection to acceptance. Our study elucidates the molecular and neural mechanisms regulating the behavioral selection process of the pre-mating female.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-grünfluoreszierendes Protein (GFP)-Antikörper in Maus hergestellte Antikörper, clone GFP-20, ascites fluid