Merck
  • Transiently "Undead" Enterocytes Mediate Homeostatic Tissue Turnover in the Adult Drosophila Midgut.
Transiently "Undead" Enterocytes Mediate Homeostatic Tissue Turnover in the Adult Drosophila Midgut.
Cell reports (2020-11-26)
Alla Amcheslavsky, Jillian L Lindblad, Andreas Bergmann
ZUSAMMENFASSUNG
We reveal surprising similarities between homeostatic cell turnover in adult Drosophila midguts and "undead" apoptosis-induced compensatory proliferation (AiP) in imaginal discs. During undead AiP, immortalized cells signal for AiP, allowing its analysis. Critical for undead AiP is the Myo1D-dependent localization of the initiator caspase Dronc to the plasma membrane. Here, we show that Myo1D functions in mature enterocytes (ECs) to control mitotic activity of intestinal stem cells (ISCs). In Myo1D mutant midguts, many signaling events involved in AiP (ROS generation, hemocyte recruitment, and JNK signaling) are affected. Importantly, similar to AiP, Myo1D is required for membrane localization of Dronc in ECs. We propose that ECs destined to die transiently enter an undead-like state through Myo1D-dependent membrane localization of Dronc, which enables them to generate signals for ISC activity and their replacement. The concept of transiently "undead" cells may be relevant for other stem cell models in flies and mammals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
ChIPAb+ Phospho-Histone H3 (Thr3) - ChIP Validated Antibody and Primer Set, rabbit monoclonal, culture supernatant, clone JY325, from rabbit