Merck
JCAD expression and localization in human blood endothelial cells.
Heliyon (2020-10-22)
Manabu Shigeoka, Satomi Arimoto, Masaya Akashi
ZUSAMMENFASSUNG
Junctional Cadherin 5 Associated (JCAD) is an endothelial, cell-cell junction protein, and its expression is associated with cardiovascular diseases including atherosclerosis and hypertension. However, to date, there are few studies confirming JCAD expression and precise localization in human tissues by immunohistochemical staining. JCAD expression and localization was assessed in four human submandibular gland (SMG) specimens by immunohistochemical staining. One specimen of SMG with sialoadenitis was accompanied by severe inflammation and fibrosis, while the other was largely normal. Other two SMGs were accompanied by severe fibrosis because of irradiation. Immunohistochemical analysis of human SMGs revealed JCAD localization at the blood endothelial cell-cell junctions. JCAD expression was more evident in microvessels and arteries in areas affected by inflammation. The localization of JCAD at endothelial cell-cell junctions was confirmed in human tissues. JCAD expression may be affected by pathological conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-KIAA1462 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution