Merck
  • Type II interferon signaling in the brain during a viral infection with age-dependent pathogenesis.
Type II interferon signaling in the brain during a viral infection with age-dependent pathogenesis.
Developmental neurobiology (2020-09-01)
Patrick S Creisher, Manisha N Chandwani, Yashika S Kamte, Jordan R Covvey, Priya Ganesan, Lauren A O'Donnell
ZUSAMMENFASSUNG
Viral infections of the central nervous system (CNS) often cause disease in an age-dependent manner, with greater neuropathology during the fetal and neonatal periods. Transgenic CD46+ mice model these age-dependent outcomes through a measles virus infection of CNS neurons. Adult CD46+ mice control viral spread and survive the infection in an interferon gamma (IFNγ)-dependent manner, whereas neonatal CD46+ mice succumb despite similar IFNγ expression in the brain. Thus, we hypothesized that IFNγ signaling in the adult brain may be more robust, potentially due to greater basal expression of IFNγ signaling proteins. To test this hypothesis, we evaluated the expression of canonical IFNγ signaling proteins in the neonatal and adult brain, including the IFNγ receptor, Janus kinase (JAK) 1/2, and signal transducer and activator of transcription-1 (STAT1) in the absence of infection. We also analyzed the expression and activation of STAT1 and IFNγ-stimulated genes during MV infection. We found that neonatal brains have equivalent or greater JAK/STAT1 expression in the hippocampus and the cerebellum than adults. IFNγ receptor expression varied by cell type in the brain but was widely expressed on neuronal and glial cells. During MV infection, increased STAT1 expression and activation correlated with viral load in the hippocampus regardless of age, but not in the cerebellum where viral load was consistently undetectable in adults. These results suggest the neonatal brain is capable of initiating IFNγ signaling during a viral infection, but that downstream STAT1 activation is insufficient to limit viral spread.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
DNAse I, grade II, from bovine pancreas
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Phosphatgepufferte Kochsalzlösung mit 0,05 % TWEEN® 20, pH 7,4
Sigma-Aldrich
L-Cystein, 97%
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Papain aus papaya latex, buffered aqueous suspension, 2× Crystallized, ≥16 units/mg protein
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN-Antikörper, serum, from guinea pig
Sigma-Aldrich
Anti-Measles Blend Antibody, hemagglutinin, clones CV1, CV4, ascites fluid, Chemicon®, from mouse
Sigma-Aldrich
Anti-Masern-Antikörper (Mischung, Matrixprotein, Klone CV8 und CV9), Chemicon®, from mouse