Merck
  • Noradrenaline signaling in the LPBN mediates amylin's and salmon calcitonin's hypophagic effect in male rats.
Noradrenaline signaling in the LPBN mediates amylin's and salmon calcitonin's hypophagic effect in male rats.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-09-29)
Lavinia Boccia, Christelle Le Foll, Thomas A Lutz
ZUSAMMENFASSUNG
The LPBN (lateral parabrachial nucleus) plays an important role in feeding control. CGRP (calcitonin gene-related peptide) LPBN neurons activation mediates the anorectic effects of different gut-derived peptides, including amylin. Amylin and its long acting analog sCT (salmon calcitonin) exert their anorectic actions primarily by directly activating neurons located in the area postrema (AP). A large proportion of projections from the AP and the adjacent nucleus of the solitary tractNTS to the LPBN, are noradrenergic (NA), and amylin-activated NAAP neurons are critical in mediating amylin's hypophagic effects. Here, we determine the functional role of NAAP amylin activated neurons to activate CGRP and non-CGRP LPBN neurons. To this end, NA was specifically depleted in the rat LPBN through a stereotaxic microinfusion of 6-OHDA, a neurotoxic agent that destroys NA terminals. While amylin (50 μg/kg) and sCT (5 μg/kg) reduced eating in sham-lesioned rats, no reduction in feeding occurred in NA-depleted animals. Further, the amylin-induced c-Fos response in the LPBN and c-Fos/CGRP colocalization were reduced in NA-depleted animals compared to controls. We conclude that AP → LPBN NA signaling, through the activation of LPBN CGRP neurons, mediates part of amylin's hypophagic effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Hydroxydopamin -hydrochlorid, ≥97% (titration), powder
Sigma-Aldrich
Pargylin -hydrochlorid
Sigma-Aldrich
Anti-Dopamin-β-Hydroxylase-Antikörper, Klon 4F10.2, ascites fluid, clone 4F10.2, Chemicon®