Merck
  • Enhanced vacuum UV-based process (VUV/H2O2/PMS) for the effective removal of ammonia from water: Engineering configuration and mechanistic considerations.
Enhanced vacuum UV-based process (VUV/H2O2/PMS) for the effective removal of ammonia from water: Engineering configuration and mechanistic considerations.
Journal of hazardous materials (2020-12-02)
Hawzhin Amanollahi, Gholamreza Moussavi, Stefanos Giannakis
ZUSAMMENFASSUNG
In this work, the VUV, VUV/H2O2, VUV/PMS, and VUV/H2O2/PMS processes were compared with the corresponding UVC-based AOPs under identical experimental conditions for the ammonia removal. Among the examined AOPs, the VUV/H2O2/PMS demonstrated the highest performance in converting NH4+ to N2. A 82.7 % removal of 100 mg/L NH4+, with N2 selectivity over 99 % was obtained in the VUV/H2O2/PMS process within 60 min, operated under near neutral pH. Under these operation conditions, [NO3-] was around 0.5 mg-N/L with [NO2-] remaining below detection. The VUV-mediated generation of SO4•-and HO• with NH4+ had a relative contribution of 37.9 and 62.1 %, respectively. The VUV/H2O2/PMS process operated under a flow-through mode achieved efficient removal of 100 mg/L NH4+ (80.5 %) in a hydraulic retention time (HRT) of 40 min. The continuous-flow VUV/H2O2/PMS process efficiently treated a real ammonia-laden groundwater and the concentration of NH4+ decreased from 30 mg/L to around 1 mg/L within 60 min HRT. In summary, the VUV/H2O2/PMS process was effective from the technical and energetical point of view, hence is a viable and promising technique for treating effluent containing high concentrations of ammonia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenazin-Methosulfat