Merck
  • Complex Autoinflammatory Syndrome Unveils Fundamental Principles of JAK1 Kinase Transcriptional and Biochemical Function.
Complex Autoinflammatory Syndrome Unveils Fundamental Principles of JAK1 Kinase Transcriptional and Biochemical Function.
Immunity (2020-08-05)
Conor N Gruber, Jorg J A Calis, Sofija Buta, Gilad Evrony, Jerome C Martin, Skyler A Uhl, Rachel Caron, Lauren Jarchin, David Dunkin, Robert Phelps, Bryn D Webb, Jeffrey M Saland, Miriam Merad, Jordan S Orange, Emily M Mace, Brad R Rosenberg, Bruce D Gelb, Dusan Bogunovic
ZUSAMMENFASSUNG
Autoinflammatory disease can result from monogenic errors of immunity. We describe a patient with early-onset multi-organ immune dysregulation resulting from a mosaic, gain-of-function mutation (S703I) in JAK1, encoding a kinase essential for signaling downstream of >25 cytokines. By custom single-cell RNA sequencing, we examine mosaicism with single-cell resolution. We find that JAK1 transcription was predominantly restricted to a single allele across different cells, introducing the concept of a mutational "transcriptotype" that differs from the genotype. Functionally, the mutation increases JAK1 activity and transactivates partnering JAKs, independent of its catalytic domain. S703I JAK1 is not only hypermorphic for cytokine signaling but also neomorphic, as it enables signaling cascades not canonically mediated by JAK1. Given these results, the patient was treated with tofacitinib, a JAK inhibitor, leading to the rapid resolution of clinical disease. These findings offer a platform for personalized medicine with the concurrent discovery of fundamental biological principles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Ionomycin Calciumsalz aus Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Anti-Phosphotyrosin-Antikörper, Klon 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-STAT2-Antikörper, CT, Upstate®, from rabbit