Merck
  • In vitro susceptibilities of 217 clinical isolates of zygomycetes to conventional and new antifungal agents.
In vitro susceptibilities of 217 clinical isolates of zygomycetes to conventional and new antifungal agents.
Antimicrobial agents and chemotherapy (2007-04-25)
Nikolaos G Almyroudis, Deanna A Sutton, Annette W Fothergill, Michael G Rinaldi, Shimon Kusne
ZUSAMMENFASSUNG
We evaluated the in vitro susceptibilities of 217 zygomycetes to amphotericin B, ketoconazole, fluconazole, itraconazole, voriconazole, posaconazole, caspofungin, and flucytosine. The significant in vitro activity of posaconazole against several species appears to support its reported clinical efficacy. Decreased susceptibility to amphotericin B was noted with Cunninghamella bertholletiae.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Fluorcytosin, nucleoside analog
Sigma-Aldrich
Ketoconazol, 99.0-101.0% (EP, titration), meets EP testing specifications