Merck
  • Rb-mediated heterochromatin formation and silencing of E2F target genes during cellular senescence.
Rb-mediated heterochromatin formation and silencing of E2F target genes during cellular senescence.
Cell (2003-06-18)
Masashi Narita, Sabrina Nũnez, Edith Heard, Masako Narita, Athena W Lin, Stephen A Hearn, David L Spector, Gregory J Hannon, Scott W Lowe
ZUSAMMENFASSUNG
Cellular senescence is an extremely stable form of cell cycle arrest that limits the proliferation of damaged cells and may act as a natural barrier to cancer progression. In this study, we describe a distinct heterochromatic structure that accumulates in senescent human fibroblasts, which we designated senescence-associated heterochromatic foci (SAHF). SAHF formation coincides with the recruitment of heterochromatin proteins and the retinoblastoma (Rb) tumor suppressor to E2F-responsive promoters and is associated with the stable repression of E2F target genes. Notably, both SAHF formation and the silencing of E2F target genes depend on the integrity of the Rb pathway and do not occur in reversibly arrested cells. These results provide a molecular explanation for the stability of the senescent state, as well as new insights into the action of Rb as a tumor suppressor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-p16 (Ab-1)-Maus-mAb (DCS-50.1/H4)-Antikörper, liquid, clone DCS-50.1/H4, Calbiochem®
Sigma-Aldrich
Anti-c-H-Ras (Ab-1) Mouse mAb (F235-1.7.1), liquid, clone F235-1.7.1, Calbiochem®