Merck
  • Antiviral and anti-inflammatory effects of rosmarinic acid in an experimental murine model of Japanese encephalitis.
Antiviral and anti-inflammatory effects of rosmarinic acid in an experimental murine model of Japanese encephalitis.
Antimicrobial agents and chemotherapy (2007-06-20)
Vivek Swarup, Joydeep Ghosh, Soumya Ghosh, Amit Saxena, Anirban Basu
ZUSAMMENFASSUNG
Rosmarinic acid (RA) reduced the mortality of mice infected with Japanese encephalitis virus (JEV). Significant decreases in viral loads (P < 0.001) and proinflammatory cytokine levels (P < 0.001) were observed in JEV-infected animals treated with RA compared to levels in infected mice without treatment, at 8 to 9 days postinfection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rosmarinsäure, 96%