Merck
  • Active N6-Methyladenine Demethylation by DMAD Regulates Gene Expression by Coordinating with Polycomb Protein in Neurons.
Active N6-Methyladenine Demethylation by DMAD Regulates Gene Expression by Coordinating with Polycomb Protein in Neurons.
Molecular cell (2018-08-07)
Bing Yao, Yujing Li, Zhiqin Wang, Li Chen, Mickael Poidevin, Can Zhang, Li Lin, Feng Wang, Han Bao, Bin Jiao, Junghwa Lim, Ying Cheng, Luoxiu Huang, Brittany Lynn Phillips, Tianlei Xu, Ranhui Duan, Kenneth H Moberg, Hao Wu, Peng Jin
ZUSAMMENFASSUNG
A ten-eleven translocation (TET) ortholog exists as a DNA N6-methyladenine (6mA) demethylase (DMAD) in Drosophila. However, the molecular roles of 6mA and DMAD remain unexplored. Through genome-wide 6mA and transcriptome profiling in Drosophila brains and neuronal cells, we found that 6mA may epigenetically regulate a group of genes involved in neurodevelopment and neuronal functions. Mechanistically, DMAD interacts with the Trithorax-related complex protein Wds to maintain active transcription by dynamically demethylating intragenic 6mA. Accumulation of 6mA by depleting DMAD coordinates with Polycomb proteins and contributes to transcriptional repression of these genes. Our findings suggest that active 6mA demethylation by DMAD plays essential roles in fly CNS by orchestrating through added epigenetic mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Anti-FLAG® M2 Magnetperlen, affinity isolated antibody
Millipore
Monoklonale Anti-HA−Agarose in Maus hergestellte Antikörper, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Lithiumchlorid, BioXtra, ≥99.0% (titration)
Millipore
EZview Red Anti-c-Myc-Affinitätsgel