Merck
FSH1 encodes lysophospholipase activity in Saccharomyces cerevisiae.
Biotechnology letters (2020-09-14)
Gowsalya Ramachandran, Ravi Chidambaram, Vasanthi Nachiappan
ZUSAMMENFASSUNG
To elucidate the role of FSH1 (family of serine hydrolase) in lipid homeostasis. Proteins in various species containing alpha/beta hydrolase domain are known to be involved in lipid metabolism. In silico analysis of the FSH1 gene in Saccharomyces cerevisiae revealed the presence of alpha/beta hydrolase domain (ABHD) and a lipase motif (GXSXG), however its function in lipid metabolism remained elusive. The overexpression of FSH1 in WT and fsh1Δ cells showed a significant reduction in the cellular phospholipid levels and an increase in the triacylglycerol levels and lipid droplet (LD) number. Furthermore, the purified recombinant protein Fsh1p was identified as a lysophospholipase that specifically acts on lysophosphatidylserine (LPS) and impacts the lipid homeostasis in S. cerevisiae. These results depicted that Fsh1p has a role on lipid homeostasis and is a lysophospholipase that hydrolyzes lysophosphatidylserine (LPS).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nitrilotriessigsäure Dinatriumsalz, Sigma Grade, ≥99%
Triolein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,2-Dioleoyl-sn-Glycerin, ≥97%