Merck
  • The PDZ domain protein PICK1 and the sodium channel BNaC1 interact and localize at mechanosensory terminals of dorsal root ganglion neurons and dendrites of central neurons.
The PDZ domain protein PICK1 and the sodium channel BNaC1 interact and localize at mechanosensory terminals of dorsal root ganglion neurons and dendrites of central neurons.
The Journal of biological chemistry (2001-12-12)
Anne Duggan, Jaime Garcia-Anoveros, David P Corey
ZUSAMMENFASSUNG
Members of the BNaC/ASIC family of ion channels have been implicated in mechanotransduction and nociception mediated by dorsal root ganglion (DRG) neurons. These ion channels are also expressed in the CNS. We identified the PDZ domain protein PICK1 as an interactor of BNaC1(ASIC2) in a yeast two-hybrid screen. We show by two-hybrid assays, glutathione S-transferase pull-down assays, and coimmunoprecipitations that the BNaC1-PICK1 interaction is specific, and that coexpression of both proteins leads to their clustering in intracellular compartments. The interaction between BNaC1 and PICK1 requires the PDZ domain of PICK1 and the last four amino acids of BNaC1. BNaC1 is similar to two other BNaC/ASIC family members, BNaC2 (ASIC1) and ASIC4, at its extreme C terminus, and we show that PICK1 also interacts with BNaC2. We found that PICK1, like BNaC1 and BNaC2, is expressed by DRG neurons and, like the BNaC1alpha isoform, is present at their peripheral mechanosensory endings. Both PICK1 and BNaC1alpha are also coexpressed by some pyramidal neurons of the cortex, by pyramidal neurons of the CA3 region of hippocampus, and by cerebellar Purkinje neurons, localizing to their dendrites and cell bodies. Therefore, PICK1 interacts with BNaC/ASIC channels and may regulate their subcellular distribution or function in both peripheral and central neurons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-c-Myc-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-muscarinischer-Acetylcholinrezeptor-m2-Antikörper, Klon M2-2-B3, clone M2-2-B3, Chemicon®, from rat