Merck
  • Surgical ablation of atrial fibrillation concomitant to coronary-artery bypass grafting provides cost-effective mortality reduction.
Surgical ablation of atrial fibrillation concomitant to coronary-artery bypass grafting provides cost-effective mortality reduction.
The Journal of thoracic and cardiovascular surgery (2019-10-16)
J Scott Rankin, Daniel J Lerner, Mary Jo Braid-Forbes, Michelle M McCrea, Vinay Badhwar
ZUSAMMENFASSUNG
Data on the longitudinal impact of surgical ablation (SA) for atrial fibrillation (AF) in patients undergoing coronary artery bypass grafting (CABG) remain limited. This study examined 2-year risk-adjusted mortality and total hospital costs in Medicare beneficiaries with AF requiring CABG with or without SA. CABG was performed in 3745 Medicare beneficiaries with AF in 2013, with concomitant SA in 17% (626 of 3745). Risk-adjusted mortality, morbidity, and cost during the first 2 postoperative years for patients with SA and those without SA were compared. A piecewise Cox proportional hazard model (0-90 days and 91-729 days) was used to risk-adjust mortality. Compared with the no SA group, the SA group had lower rates of heart failure before surgery (31% vs 36%), chronic lung disease (27% vs 33%), renal failure (4% vs 7%), and urgent or emergent presentation (34% vs 49%) (all P < .05). Risk-adjusted index admission costs were higher with SA (rate ratio [RR], 1.11; P < .01), as were readmissions for AF (hazard ratio [HR], 1.14; 95% confidence interval [CI], 1.00-1.29; P = .04) and pacemaker/defibrillator implantation (HR, 1.37; 95%, 1.08-1.74; P = .01). Risk-adjusted inpatient days and inpatient costs were similar after 2 years (RR, 0.97; P = .31 and RR = 1.04; P = .17, respectively); however, the risk-adjusted hazard for late mortality (91-729 days) was significantly lower with SA (HR, 0.71; 95% CI, 0.52-0.97; P = .03). In patients with AF requiring CABG, SA was associated with a 29% lower risk-adjusted hazard for late mortality. Index hospital costs were higher with SA, but total inpatient costs were not different in the 2 groups after 2 years. SA appears to be a cost-effective intervention to enhance late 2-year survival in patients with AF undergoing CABG.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder