Merck
  • The transcriptional regulator Zfat is essential for maintenance and differentiation of the adipocytes.
The transcriptional regulator Zfat is essential for maintenance and differentiation of the adipocytes.
Journal of cellular biochemistry (2021-02-02)
Shuhei Ishikura, Masayoshi Nagai, Toshiyuki Tsunoda, Kensuke Nishi, Yoko Tanaka, Midori Koyanagi, Senji Shirasawa
ZUSAMMENFASSUNG
Adipocytes play crucial roles in the control of whole-body energy homeostasis. Differentiation and functions of the adipocytes are regulated by various transcription factors. Zfat (zinc-finger protein with AT-hook) is a transcriptional regulator that controls messenger RNA expression of specific genes through binding to their transcription start sites. Here we report important roles of Zfat in the adipocytes. We establish inducible Zfat-knockout (Zfat iKO) mice where treatment with tamoxifen causes a marked reduction in Zfat expression in various tissues. Tamoxifen treatment of Zfat iKO mice reduces the white adipose tissues (WATs) mass, accompanied by the decreased triglyceride levels. Zfat is expressed in both the adipose-derived stem cells (ADSCs) and mature adipocytes in the WATs. In ex vivo assays of the mature adipocytes differentiated from the Zfat iKO ADSCs, loss of Zfat in the mature adipocytes reduces the triglyceride levels, suggesting cell autonomous roles of Zfat in the maintenance of the mature adipocytes. Furthermore, we identify the Atg13, Brf1, Psmc3, and Timm22 genes as Zfat-target genes in the mature adipocytes. In contrast, loss of Zfat in the ADSCs impairs adipocyte differentiation with the decreased expression of C/EBPα and adiponectin. Thus, we propose that Zfat plays crucial roles in maintenance and differentiation of the adipocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Roche
Dispase® II (neutrale Protease, Klasse II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution