Merck
SRF Is Required for Maintenance of Astrocytes in Non-Reactive State in the Mammalian Brain.
eNeuro (2021-01-15)
Monika Jain, Soumen Das, Paul P Y Lu, Garima Virmani, Sumitha Soman, Surya Chandra Rao Thumu, David H Gutmann, Narendrakumar Ramanan
ZUSAMMENFASSUNG
Astrocytes play several critical roles in the normal functioning of the mammalian brain, including ion homeostasis, synapse formation, and synaptic plasticity. Following injury and infection or in the setting of neurodegeneration, astrocytes become hypertrophic and reactive, a process termed astrogliosis. Although acute reactive gliosis is beneficial in limiting further tissue damage, chronic gliosis becomes detrimental for neuronal recovery and regeneration. Several extracellular factors have been identified that generate reactive astrocytes; however, very little is known about the cell-autonomous transcriptional mechanisms that regulate the maintenance of astrocytes in the normal non-reactive state. Here, we show that conditional deletion of the stimulus-dependent transcription factor, serum response factor (SRF) in astrocytes (SrfGFAPCKO) results in astrogliosis marked by hypertrophic morphology and increased expression of GFAP, vimentin, and nestin. These reactive astrocytes were not restricted to any specific brain region and were seen in both white and gray matter in the entire brain. This astrogliosis persisted throughout adulthood concomitant with microglial activation. Importantly, the Srf mutant mouse brain did not exhibit any cell death or blood brain barrier (BBB) deficits suggesting that apoptosis and leaky BBB are not the causes for the reactive phenotype. The mutant astrocytes expressed more A2 reactive astrocyte marker genes and the SrfGFAPCKO mice exhibited normal neuronal numbers indicating that SRF-deficient gliosis astrocytes are not neurotoxic. Together, our findings suggest that SRF plays a critical role in astrocytes to maintain them in a non-reactive state.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoklonaler Anti-GFAP-Antikörper (Glial Fibrillary Acidic Protein, Saures Gliafaserprotein) in Maus hergestellte Antikörper, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Nestin-Antikörper, Klon 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Anti-S-100 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-Histone H3 (pSer10) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution