Merck
  • Analysis of segmentation ontology reveals the similarities and differences in connectivity onto L2/3 neurons in mouse V1.
Analysis of segmentation ontology reveals the similarities and differences in connectivity onto L2/3 neurons in mouse V1.
Scientific reports (2021-03-04)
Alexander P Y Brown, Lee Cossell, Molly Strom, Adam L Tyson, Mateo Vélez-Fort, Troy W Margrie
ZUSAMMENFASSUNG
Quantitatively comparing brain-wide connectivity of different types of neuron is of vital importance in understanding the function of the mammalian cortex. Here we have designed an analytical approach to examine and compare datasets from hierarchical segmentation ontologies, and applied it to long-range presynaptic connectivity onto excitatory and inhibitory neurons, mainly located in layer 2/3 (L2/3), of mouse primary visual cortex (V1). We find that the origins of long-range connections onto these two general cell classes-as well as their proportions-are quite similar, in contrast to the inputs on to a cell type in L6. These anatomical data suggest that distal inputs received by the general excitatory and inhibitory classes of neuron in L2/3 overlap considerably.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoclonal Anti-GABA antibody produced in mouse, clone GB-69, ascites fluid