Merck
  • Synthesis and anticancer activity comparison of phenylalkyl isoselenocyanates with corresponding naturally occurring and synthetic isothiocyanates.
Synthesis and anticancer activity comparison of phenylalkyl isoselenocyanates with corresponding naturally occurring and synthetic isothiocyanates.
Journal of medicinal chemistry (2008-12-05)
Arun K Sharma, Arati Sharma, Dhimant Desai, SubbaRao V Madhunapantula, Sung Jin Huh, Gavin P Robertson, Shantu Amin
ZUSAMMENFASSUNG
Synthesis and identification of novel phenylalkyl isoselenocyanates (ISCs), isosteric selenium analogues of naturally occurring phenylalkyl isothiocyanates (ITCs), as effective cytotoxic and antitumor agents are described. The structure-activity relationship comparison of ISCs with ITCs and effect of the increasing alkyl chain length in inhibiting cancer cell growth were evaluated on melanoma, prostate, breast, glioblastoma, sarcoma, and colon cancer cell lines. IC(50) values for ISC compounds were generally lower than their corresponding ITC analogues. Similarly, in UACC 903 human melanoma cells, the inhibition of cell proliferation and induction of apoptosis were more pronounced with ISCs compared to ITCs. Further, ISCs and ITCs effectively inhibited melanoma tumor growth in mice following intraperitoneal xenograft. A similar reduction in tumor size was observed at 3 times lower doses of ISCs compared to corresponding ITCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzylisothiocyanat, 98%
Sigma-Aldrich
Phenethylisothiocyanat, 99%