Merck
AlbuCORE: an albumin-based molecular scaffold for multivalent biologics design.
mAbs (2020-08-21)
Mario Sanches, Igor D'Angelo, Maria Jaramillo, Jason Baardsnes, John Zwaagstra, Joe Schrag, Ian Schoenhofen, Mauro Acchione, Sam Lawn, Grant Wickman, Nina Weisser, David K Y Poon, Gordon Ng, Surjit Dixit
ZUSAMMENFASSUNG
As biologics have become a mainstay in the development of novel therapies, protein engineering tools to expand on their structural advantages, namely specificity, affinity, and valency are of interest. Antibodies have dominated this field as the preferred scaffold for biologics development while there has been limited exploration into the use of albumin with its unique physiological characteristics as a platform for biologics design. There has been a great deal of interest to create bispecific and more complex multivalent molecules to build on the advantages offered by protein-based therapeutics relative to small molecules. Here, we explore the use of human serum albumin (HSA) as a scaffold for the design of multispecific biologics. In particular, we describe a structure-guided approach to the design of split HSA molecules we refer to as AlbuCORE, that effectively and spontaneously forms a native albumin-like molecule, but in a heterodimeric state upon co-expression. We show that the split AlbuCORE designs allow the creation of novel fusion entities with unique alternate geometries. We also show that, apart from these AlbuCORE fusion entities, there is an opportunity to explore their albumin-like small hydrophobic molecule carrying capacity as a drug conjugate in these designs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin human, Cellastim S, recombinant, expressed in rice, lyophilized powder, suitable for cell culture, low endotoxin, ≥96% (SDS-PAGE)
Sigma-Aldrich
Albumin aus Humanserum, lyophilized powder, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-Albumin antibody produced in goat, fractionated antiserum, lyophilized