Merck
Discovery of amino-acetonitrile derivatives, a new class of synthetic anthelmintic compounds.
Bioorganic & medicinal chemistry letters (2008-04-11)
Pierre Ducray, Noëlle Gauvry, François Pautrat, Thomas Goebel, Joerg Fruechtel, Yves Desaules, Sandra Schorderet Weber, Jacques Bouvier, Trixie Wagner, Olivier Froelich, Ronald Kaminsky
ZUSAMMENFASSUNG
A new series of amino-acetonitrile derivatives (AAD) have been discovered that exhibit high anthelmintic activity against parasitic nematode species such as Haemonchus contortus and Trichostrongylus colubriformis. Significantly, these compounds also demonstrate activity against nematode strains resistant to the currently available broad-spectrum anthelmintics. The discovery, synthesis, structure-activity relationship and biological results are presented.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiabendazol, ≥99%, powder
Sigma-Aldrich
Thiabendazol, BioReagent, suitable for plant cell culture, powder
Supelco
Thiabendazol, PESTANAL®, analytical standard