Merck
From thymus to cystic fibrosis: the amazing life of thymosin alpha 1.
Expert opinion on biological therapy (2018-08-01)
Enrico Garaci
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thymosin α1 bovine, ≥90% (HPLC)