Merck
  • Higher Anti-Tumor Efficacy of the Dual HER3-EGFR Antibody MEHD7945a Combined with Ionizing Irradiation in Cervical Cancer Cells.
Higher Anti-Tumor Efficacy of the Dual HER3-EGFR Antibody MEHD7945a Combined with Ionizing Irradiation in Cervical Cancer Cells.
International journal of radiation oncology, biology, physics (2020-01-22)
Laura Bourillon, Sylvain Demontoy, Alexis Lenglet, Alexandre Zampieri, Julien Fraisse, Marta Jarlier, Florence Boissière-Michot, Hélène Perrochia, Gauthier Rathat, Véronique Garambois, Nathalie Bonnefoy, Henri-Alexandre Michaud, Thierry Chardès, Diego Tosi, André Pèlegrin, David Azria, Christel Larbouret, Céline Bourgier
ZUSAMMENFASSUNG
The outcome of locally advanced cervical cancer (LACC) is dismal. Biomarkers are needed to individualize treatments and to improve patient outcomes. Here, we investigated whether coexpression of epidermal growth factor receptor (EGFR) and human epidermal growth factor receptor 3 (HER3) could be an outcome prognostic biomarker, and whether targeting both EGFR and HER3 with a dual antibody (MEHD7945A) enhanced ionizing radiation (IR) efficacy. Expression of EGFR and HER3 was evaluated by immunohistochemistry in cancer biopsies (n = 72 patients with LACC). The antitumor effects of the MEHD7945A and IR combotherapy were assessed in 2 EGFR- and HER3-positive cervical cancer cell lines (A431 and CaSki) and in A431 cell xenografts. The mechanisms involved in tumor cell radiosensitization were also studied. The interaction of MEHD7945A, IR, and cisplatin was evaluated using dose-response matrix data. EGFR and HER3 were coexpressed in only in 7 of the 22 biopsies of FIGO IVB cervix cancer. The median overall survival was 14.6 months and 23.1 months in patients with FIGO IVB tumors that coexpressed or did not coexpress EGFR and HER3, respectively. In mice xenografted with A431 (squamous cell carcinoma) cells, MEHD7945A significantly increased IR response by reducing tumor growth and increasing cleaved caspase-3 expression. In A431 and CaSki cells, the combotherapy increased DNA damage and cell death, particularly immunogenic cell death, and decreased survival by inhibiting the MAPK and AKT pathways. An additive effect was observed when IR, MEHD7945A, and cisplatin were combined. Targeting EGFR and HER3 with a specific dual antibody enhanced IR efficacy. These preliminary results and the prognostic value of EGFR and HER3 coexpression should be confirmed in a larger sample.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-phospho-Histon H2A.X (Ser139)-Antikörper, Klon JBW301, clone JBW301, Upstate®, from mouse