Merck
Coactosin-Like Protein (COTL1) Promotes Glioblastoma (GBM) Growth in vitro and in vivo.
Cancer management and research (2020-11-07)
Shike Shao, Yongjun Fan, Chongpei Zhong, Xianlong Zhu, Jiaqiu Zhu
ZUSAMMENFASSUNG
To assess the expression levels of COTL1 in human GBM tissues and evaluate the potential involvement of COTL1 in cancer progression. Bioinformation analysis was performed to evaluate COTL1 mRNA levels in GBM tissues and normal tissues, according to the TCGA database, and explore the effects on prognosis. Immunohistochemical (IHC) assays were performed to evaluate COTL1 expression in human GBM tissues and the clinical pathological analysis was performed. Colony formation and MTT assays were performed to evaluate the effects of COTL1 on GBM cell proliferation. Immunoblot assays were performed to detect the expression level of COTL1, Ki67, and PCNA. A xenograft model was developed in mice to assess the effects of COTL1 on tumor growth in vivo. We found COTL1 had an obvious high expression in human GBM tissues. The expression of COTL1 was related to recurrence (P=0.006**) and prognosis of patients with GBM. Our data further demonstrated COTL1 promoted cell proliferation in vitro and contributed to tumor growth of GBM cells in mice. We therefore identified a novel and promising therapeutic target for the treatment of GBM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-PCNA, IgG fraction of antiserum
Sigma-Aldrich
MISSION® esiRNA, targeting human COTL1