Merck
  • A Bispecific Single-Domain Antibody Boosts Autologous Vγ9Vδ2-T Cell Responses Toward CD1d in Chronic Lymphocytic Leukemia.
A Bispecific Single-Domain Antibody Boosts Autologous Vγ9Vδ2-T Cell Responses Toward CD1d in Chronic Lymphocytic Leukemia.
Clinical cancer research : an official journal of the American Association for Cancer Research (2021-01-17)
Iris de Weerdt, Roeland Lameris, Jurjen M Ruben, Renate de Boer, Jan Kloosterman, Lisa A King, Mark-David Levin, Paul W H I Parren, Tanja D de Gruijl, Arnon P Kater, Hans J van der Vliet
ZUSAMMENFASSUNG
Although considerable progress has been made with autologous T cell-based therapy in B-cell malignancies, application in chronic lymphocytic leukemia (CLL) lags behind due to disappointing response rates as well as substantial toxicity that is of particular concern in the elderly CLL population. Vγ9Vδ2-T cells form a conserved T-cell subset with strong intrinsic immunotherapeutic potential, largely because of their capacity to be triggered by phosphoantigens that can be overproduced by CLL and other malignant cells. Specific activation of Vγ9Vδ2-T cells by a bispecific antibody may improve the efficacy and toxicity of autologous T-cell-based therapy in CLL. We evaluated CD1d expression in a cohort of 78 untreated patients with CLL and generated and functionally characterized a CD1d-specific Vγ9Vδ2-T cell engager based on single-domain antibodies (VHH). CD1d was expressed by CLL in the majority of patients, particularly in patients with advanced disease. The CD1d-specific Vγ9Vδ2-T cell engager induced robust activation and degranulation of Vγ9Vδ2-T cells, enabling Vγ9Vδ2-T cells from patients with CLL to lyse autologous leukemic cells at low effector-to-target ratios. Expression of CD1d on CLL cells is upregulated by all-trans retinoic acid, and sensitizes the malignant cells to bispecific VHH-induced lysis. Furthermore, we provide evidence that the Vγ9Vδ2-T cell receptor retains responsiveness to phosphoantigens when the bispecific VHH is bound, and aminobisphosphonates can therefore enhance bispecific Vγ9Vδ2-T cell engager-mediated tumor-specific killing. Collectively, our data demonstrate the immunotherapeutic potential of this novel CD1d-specific Vγ9Vδ2-T cell engager in CLL.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fetales Kälberserum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Mevastatin, ≥98% (HPLC), powder or crystals