Merck
  • Carbonic anhydrase activators: activation of the human tumor-associated isozymes IX and XII with amino acids and amines.
Carbonic anhydrase activators: activation of the human tumor-associated isozymes IX and XII with amino acids and amines.
Bioorganic & medicinal chemistry (2008-02-26)
Silvia Pastorekova, Daniela Vullo, Isao Nishimori, Andrea Scozzafava, Jaromir Pastorek, Claudiu T Supuran
ZUSAMMENFASSUNG
The first activation study of the human carbonic anhydrase (hCA, EC 4.2.1.1) isoforms associated to tumors, hCA IX and XII, with a small library of natural and non-natural amino acids as well as aromatic/heterocyclic amines is reported. hCA IX was activated efficiently by dopamine, adrenaline and heterocyclic amines possessing aminoethyl-/aminomethyl-moieties (K(A)s of 9 nM-1.07 microM), whereas the best hCA XII activators were serotonin, L-adrenaline, 4-(2-aminoethyl)-morpholine and d-Phe (K(A) of 0.24-0.41 microM). Precise steric and electronic requirements are needed to be present in the molecules of effective hCA IX/hCA XII activators, in order to assure an adequate fit within the enzyme active site cavity for the formation of the enzyme-activator complex, and for an efficient proton transfer process within this complex, leading to the release of a proton and formation of the catalytically active, zinc-hydroxide species of the enzyme. Selective activation of these CA isoforms might be useful to develop pharmacologic tools or to understand whether some of these biogenic amines/amino acids may influence the progression of tumors overexpressing CA IX and/or CA XII.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Histidin, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
L-Tyrosin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Histidin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidin, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Phenylalanin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
SAFC
L-Histidin
Sigma-Aldrich
Histamin, ≥97.0%
Sigma-Aldrich
D-Phenylalanin, ≥98% (HPLC)
Sigma-Aldrich
L-Phenylalanin, reagent grade, ≥98%
Sigma-Aldrich
2-Picolylamin, 99%
Sigma-Aldrich
1-(2-Aminoethyl)piperazin, 99%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Phenylalanin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
4-(2-Aminoethyl)-morpholin, 99%
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
SAFC
L-Tyrosin
Sigma-Aldrich
3,4-Dihydroxy-D-Phenylalanin, powder, ≥95%
Sigma-Aldrich
D-Histidin, ≥98% (TLC)
Sigma-Aldrich
2-(2-Pyridyl)ethylamin, 95%
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosin, FG
Sigma-Aldrich
L-Phenylalanin, 99%, FCC
Supelco
L-Phenylalanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland