- Immobilization of alkaline phosphatase on magnetic particles by site-specific and covalent cross-linking catalyzed by microbial transglutaminase.
Immobilization of alkaline phosphatase on magnetic particles by site-specific and covalent cross-linking catalyzed by microbial transglutaminase.
Journal of bioscience and bioengineering (2011-03-15)
Kousuke Moriyama, Kyunga Sung, Masahiro Goto, Noriho Kamiya
PMID21398176
ZUSAMMENFASSUNG
Bacterial alkaline phosphatase (BAP) was site-specifically and covalently immobilized on magnetic particles (MPs) using the enzymatic reaction of microbial transglutaminase (MTG). Immobilization efficiency was affected by the chemical surface treatment of MPs and immobilized BAP exhibited more than 90% of the initial activity after 10 rounds of recycling.