Merck
  • Labeling primary amine groups in peptides and proteins with N-hydroxysuccinimidyl ester modified Fe3O4@SiO2 nanoparticles containing cleavable disulfide-bond linkers.
Labeling primary amine groups in peptides and proteins with N-hydroxysuccinimidyl ester modified Fe3O4@SiO2 nanoparticles containing cleavable disulfide-bond linkers.
Bioconjugate chemistry (2013-08-06)
Ujwal S Patil, Haiou Qu, Daniela Caruntu, Charles J O'Connor, Arjun Sharma, Yang Cai, Matthew A Tarr
ZUSAMMENFASSUNG
The surface of superparamagnetic silica coated iron oxide (Fe3O4@SiO2) nanoparticles was functionalized with a disulfide bond linked N-hydroxysuccinimidyl (NHS) ester group in order to develop a method for labeling primary amines in peptides/proteins. The nanoparticle labeled proteins/peptides formed after NHS ester reaction with the primary amine groups were isolated using a magnet without any additional purification step. Nanoparticle moieties conjugated to peptides/proteins were then trimmed by cleavage at the disulfide linker with a reducing agent. The labeled peptides were analyzed by LC-MS/MS to determine their sequences and the sites of NHS ester labeling. This novel approach allowed characterization of lysine residues on the solvent accessible surface of native bovine serum albumin. Low cost, rapid magnetic separation, and specificity toward primary amine groups make NHS ester coated Fe3O4@SiO2 nanoparticles a potential labeling probe to study proteins on living cell surfaces.