Merck
  • Otubain 2 is a novel promoter of beta cell survival as revealed by siRNA high-throughput screens of human pancreatic islets.
Otubain 2 is a novel promoter of beta cell survival as revealed by siRNA high-throughput screens of human pancreatic islets.
Diabetologia (2013-03-22)
A Beck, Y Vinik, H Shatz-Azoulay, R Isaac, S Streim, G Jona, S Boura-Halfon, Y Zick
ZUSAMMENFASSUNG
Pro-inflammatory cytokines induce death of beta cells and hamper engraftment of transplanted islet mass. Our aim was to reveal novel genes involved in this process, as a platform for innovative therapeutic approaches. Small interfering RNA (siRNA) high-throughput screening (HTS) of primary human islets was employed to identify novel genes involved in cytokine-induced beta cell apoptosis. Dispersed human islets from nine human donors, treated with a combination of TNF-α, IL-1β and IFN-γ were transfected with ∼730 different siRNAs. Caspase-3/7 activity was measured, results were analysed and potential anti- and pro-apoptotic genes were identified. Dispersed human pancreatic islets appeared to be suitable targets for performance of siRNA HTS. Using this methodology we found a number of potential pro- and anti-apoptotic target hits that have not been previously associated with pancreatic beta cell death. One such hit was the de-ubiquitinating enzyme otubain 2 (OTUB2). OTUB2 knockdown increased caspase-3/7 activity in MIN6 cells and primary human islets and inhibited insulin secretion and increased nuclear factor-κB (NF-κB) activity both under basal conditions and following cytokine treatment. Use of dispersed human islets provides a new platform for functional HTS in a highly physiological system. Employing this technique enabled the identification of OTUB2 as a novel promoter of viability and insulin secretion in human beta cells. OTUB2 acts through the inhibition of NF-κB signalling, which is deleterious to beta cell survival. siRNA screens of human islets may therefore identify new targets, such as OTUB2, for therapeutic intervention in type 1 diabetes and islet transplantation.