Merck
Coumestrol suppresses proliferation of ES2 human epithelial ovarian cancer cells.
The Journal of endocrinology (2015-12-25)
Whasun Lim, Wooyoung Jeong, Gwonhwa Song
ZUSAMMENFASSUNG
Coumestrol, which is predominantly found in soybean products as a phytoestrogen, has cancer preventive activities in estrogen-responsive carcinomas. However, effects and molecular targets of coumestrol have not been reported for epithelial ovarian cancer (EOC). In the present study, we demonstrated that coumestrol inhibited viability and invasion and induced apoptosis of ES2 (clear cell-/serous carcinoma origin) cells. In addition, immunoreactive PCNA and ERBB2, markers of proliferation of ovarian carcinoma, were attenuated in their expression in coumestrol-induced death of ES2 cells. Phosphorylation of AKT, p70S6K, ERK1/2, JNK1/2, and p90RSK was inactivated by coumestrol treatment in a dose- and time-dependent manner as determined in western blot analyses. Moreover, PI3K inhibitors enhanced effects of coumestrol to decrease phosphorylation of AKT, p70S6K, S6, and ERK1/2. Furthermore, coumestrol has strong cancer preventive effects as compared to other conventional chemotherapeutics on proliferation of ES2 cells. In conclusion, coumestrol exerts chemotherapeutic effects via PI3K and ERK1/2 MAPK pathways and is a potentially novel treatment regimen with enhanced chemoprevention activities against progression of EOC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Zellproliferation ELISA, BrdU (kolorimetrisch), sufficient for ≤1,000 tests
Sigma-Aldrich
Harris-Hämatoxylin-Lösung, modifiziert