Merck
  • Long non-coding RNA CCAT1 is a prognostic biomarker for the progression of oral squamous cell carcinoma via miR-181a-mediated Wnt/β-catenin signaling pathway.
Long non-coding RNA CCAT1 is a prognostic biomarker for the progression of oral squamous cell carcinoma via miR-181a-mediated Wnt/β-catenin signaling pathway.
Cell cycle (Georgetown, Tex.) (2019-10-11)
Guang-Hui Li, Zhong-Hui Ma, Xi Wang
ZUSAMMENFASSUNG
Oral squamous cell carcinoma (OSCC) ranks as the sixth most common carcinoma worldwide, and the third most common carcinoma in developing countries as well. Recently, the aberrant expression of lncRNA CCAT1 has been revealed to play an important role in the development of several cancers. However, its role in OSCC remains unknown. The expression levels of CCAT1 and miR-181a were determined in 15 paired primary OSCC tissues and their adjacent noncancerous tissues and cell lines with qPCR. shRNA against CCAT1 was employed to investigate the impact of CCAT1 on proliferation and metastasis. Then dual luciferase reporter and RIP assays were utilized to study the interaction between CCAT1 and miR-181a. Cells transfected with sh-CCAT1 or treated with miR-181a inhibitor were subjected to western blot to investigate the role of Wnt/β-catenin signaling in CCAT1-mediated proliferation and metastasis. Finally, the role of CCAT1 in OSCC was confirmed with tumor xenografts mice model. CCAT1 was upregulated in OSCC tissues and cell lines. Knockdown of CCAT1 inhibited the proliferation, migration and invasion of OSCC cells, while the cell apoptosis was enhanced. Luciferase and RIP assays revealed that miR-181a was a direct target of CCAT1. Inhibition of miR-181a partially reversed the efficacy of sh-CCAT1. Moreover, sh-CCAT1 inhibited OSCC tissues growth through inhibiting Wnt signaling in a miR-181a-dependent manner in vivo. lncRNA CCAT1 activated Wnt/β-catenin signaling via inhibiting miR-181a, resulting in the cell proliferation, migration and invasion of OSCC, suggesting that CCAT1 might serve as a potential target of OSCC treatment. Abbreviation: LncRNA: long non-coding RNA; OSCC: oral squamous cell carcinoma; 3' UTR: 3' untranslated region; ANOVA: one-way analysis of variance; CDK: cyclin-dependent kinase; ceRNA: competing endogenous RNA; FBS: fetal bovine serum; HGF: human gingival fibroblasts; MAPK: mitogen-activated protein kinase; miRNA: micro RNA; ncRNA: noncoding RNAs; PBS: phosphate-buffered saline; PI3K: phosphatidylinositol 3-kinase.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Imprint® RNA Immunoprecipitation Kit, High-capacity Protein A magnetic beads for successful RNA Immunoprecipitation,suitable for use with mRNA and microRNA