Merck
  • Lipid-lowering effects of anti-angiopoietin-like 4 antibody recapitulate the lipid phenotype found in angiopoietin-like 4 knockout mice.
Lipid-lowering effects of anti-angiopoietin-like 4 antibody recapitulate the lipid phenotype found in angiopoietin-like 4 knockout mice.
Proceedings of the National Academy of Sciences of the United States of America (2007-07-05)
Urvi Desai, E-Chiang Lee, Kyu Chung, Cuihua Gao, Jason Gay, Billie Key, Gwenn Hansen, Dennis Machajewski, Kenneth A Platt, Arthur T Sands, Matthias Schneider, Isaac Van Sligtenhorst, Adisak Suwanichkul, Peter Vogel, Nat Wilganowski, June Wingert, Brian P Zambrowicz, Greg Landes, David R Powell
ZUSAMMENFASSUNG
We used gene knockout mice to explore the role of Angiopoietin-like-4 (Angptl4) in lipid metabolism as well as to generate anti-Angptl4 mAbs with pharmacological activity. Angptl4 -/- mice had lower triglyceride (TG) levels resulting both from increased very low-density lipoprotein (VLDL) clearance and decreased VLDL production and had modestly lower cholesterol levels. Also, both Angptl4 -/- suckling mice and adult mice fed a high-fat diet showed reduced viability associated with lipogranulomatous lesions of the intestines and their draining lymphatics and mesenteric lymph nodes. Treating C57BL/6J, ApoE -/-, LDLr -/-, and db/db mice with the anti-Angptl4 mAb 14D12 recapitulated the lipid and histopathologic phenotypes noted in Angptl4 -/- mice. This demonstrates that the knockout phenotype reflects not only the physiologic function of the Angptl4 gene but also predicts the pharmacologic consequences of Angptl4 protein inhibition with a neutralizing antibody in relevant models of human disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kit zum Nachweis von Triglycerid in Serum, 1 kit sufficient for 250 tests