Merck
  • NO-donors. Part 17: Synthesis and antimicrobial activity of novel ketoconazole-NO-donor hybrid compounds.
NO-donors. Part 17: Synthesis and antimicrobial activity of novel ketoconazole-NO-donor hybrid compounds.
Bioorganic & medicinal chemistry (2008-08-20)
Joerg Konter, Ute Möllmann, Jochen Lehmann
ZUSAMMENFASSUNG
Novel hybrid compounds combining the antifungal drug ketoconazole with a diazen-1-ium-1,2-diolate or an organic nitrate moiety and the corresponding NO-donors without ketoconazole were synthesized and their activities against a broad variety of fungal strains were tested. Hybridization modifies the spectrum of antimicrobial activities and generally, the ketoconazole-NO-donor hybrids are more potent than ketoconazole. The NO-donors alone show insufficient effectiveness.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ketoconazol, 99.0-101.0% (EP, titration), meets EP testing specifications