Merck
QSAR modeling of the interaction of flavonoids with GABA(A) receptor.
European journal of medicinal chemistry (2007-12-26)
Pablo R Duchowicz, Martín G Vitale, Eduardo A Castro, Juan C Autino, Gustavo P Romanelli, Daniel O Bennardi
ZUSAMMENFASSUNG
Experimentally assigned values to binding affinity constants of flavonoid ligands towards the benzodiazepine site of the GABA(A) receptor complex were compiled from several publications, and enabled to perform a predictive analysis based on Quantitative Structure-Activity Relationships (QSAR). The best linear model established on 78 molecular structures incorporated four molecular descriptors, selected from more than a thousand of geometrical, topological, quantum-mechanical and electronic types of descriptors and calculated by Dragon software. An application of this QSAR equation was performed by estimating the binding affinities for some newly synthesized flavonoids displaying 2-,7-substitutions in the benzopyrane backbone which still do not have experimentally measured potencies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Chrysin, ≥96.5%
Sigma-Aldrich
Flavon, ≥99.0%