Merck
Muscle glycogen content modifies SR Ca2+ release rate in elite endurance athletes.
Medicine and science in sports and exercise (2013-10-05)
Kasper Degn Gejl, Lars Grøndahl Hvid, Ulrik Frandsen, Kurt Jensen, Kent Sahlin, Niels Ørtenblad
ZUSAMMENFASSUNG
The aim of the present study was to investigate the influence of muscle glycogen content on sarcoplasmic reticulum (SR) function and peak power output (Wpeak) in elite endurance athletes. Fourteen highly trained male triathletes (VO2max = 66.5 ± 1.3 mL O2·kg·min), performed 4 h of glycogen-depleting cycling exercise (HRmean = 73% ± 1% of maximum). During the first 4 h of recovery, athletes received either water (H2O) or carbohydrate (CHO), separating alterations in muscle glycogen content from acute changes affecting SR function and performance. Thereafter, all subjects received CHO-enriched food for the remaining 20-h recovery period. Immediately after exercise, muscle glycogen content and SR Ca release rate was reduced to 32% ± 4% (225 ± 28 mmol·kg dw) and 86% ± 2% of initial levels, respectively (P < 0.01). Glycogen markedly recovered after 4 h of recovery with CHO (61% ± 2% of preexercise) and SR Ca release rate returned to preexercise level. However, in the absence of CHO during the first 4 h of recovery, glycogen and SR Ca release rate remained depressed, with the normalization of both parameters at the end of the 24 h of recovery after receiving a CHO-enriched diet. Linear regression demonstrated a significant correlation between SR Ca release rate and muscle glycogen content (P < 0.01, r = 0.30). The 4 h of cycling exercise reduced Wpeak by 5.5%-8.9% at different cadences (P < 0.05), and Wpeak was normalized after 4 h of recovery with CHO, whereas Wpeak remained depressed (P < 0.05) after water provision. Wpeak was fully recovered after 24 h in both the H2O and the CHO group. In conclusion, the present results suggest that low muscle glycogen depresses muscle SR Ca release rate, which may contribute to fatigue and delayed recovery of Wpeak 4 h postexercise.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Myosin-Antikörper (skelettal, Fast) in Maus hergestellte Antikörper, clone MY-32, ascites fluid