Merck
  • Novel keto-enamine Schiffs bases from 7-hydroxy-4-methyl-2-oxo-2H-benzo[h] chromene-8,10-dicarbaldehyde as potential antidyslipidemic and antioxidant agents.
Novel keto-enamine Schiffs bases from 7-hydroxy-4-methyl-2-oxo-2H-benzo[h] chromene-8,10-dicarbaldehyde as potential antidyslipidemic and antioxidant agents.
European journal of medicinal chemistry (2007-12-08)
Koneni V Sashidhara, Jammikuntla N Rosaiah, Gitika Bhatia, J K Saxena
ZUSAMMENFASSUNG
A series of Schiff bases have been synthesized from dicarbaldehyde of benzocoumarin, in which the reactions were regioselective and the products existed in the keto-enamine form, in which the aromaticity of the relevant ring was disrupted. The compounds were evaluated in vitro for their antioxidant and in vivo for their antidyslipidemic activity for the first time. Compounds 3 and 7 possess significant lipid lowering and antioxidant activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination