Merck
  • Nutrient-stimulated glucagon-like peptide 1 release after body-weight loss and weight maintenance in human subjects.
Nutrient-stimulated glucagon-like peptide 1 release after body-weight loss and weight maintenance in human subjects.
The British journal of nutrition (2006-01-31)
Tanja C M Adam, Manuela P G M Lejeune, Margriet S Westerterp-Plantenga
ZUSAMMENFASSUNG
Glucagon-like peptide 1 (GLP-1) is a peptide hormone that is released in response to nutrient ingestion. Postprandial GLP-1 release has been reported to be attenuated in obese subjects, but reports on the effect of weight loss on GLP-1 are conflicting. The aim of the present study was to clarify the effect of a weight-loss period and a consecutive weight-maintenance period on nutrient-stimulated GLP-1 release in obese subjects. Nutrient-stimulated (standard breakfast; 1.9 MJ) GLP-1 release was investigated in thirty-two obese subjects on three occasions: before weight loss (T1) (BMI 30.0 (sd 2.5) kg/m(2)); after a 6-week very-low-energy diet (VLED) (T2) (BMI 27.6 (sd 2.3) kg/m(2)); after a 3-month weight-maintenance period (T3) (BMI 27.9 (sd 2.3) kg/m(2)). At each occasion, following a fasting blood sample the test meal was fed and blood was drawn every 30 min for 2 h relative to ingestion in order to determine plasma GLP-1, insulin, glucose and NEFA concentrations. Subjects lost 7 (sd 3.4) kg during the VLED (P<0.0001) and regained 1 (sd 3.2) kg during the weight-maintenance period (NS). The area under the curve for nutrient-stimulated plasma GLP-1 (pmol/l x h) was significantly decreased (P=0.01) at T2 (6.8 (sd 1)) compared with T1 (12.8 (sd 2.9)) and T3 (11.1 (sd 1.5)). Since we found a rebound of concentrations after a weight-maintenance period, decrease after weight loss seems to be transient and possibly due to a negative energy balance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Glucagon-like Peptide 1 (aktiv)-ELISA, This Glucagon Like Peptide-1 (Active) ELISA is used to measure & quantify Glucagon Like Peptide-1 levels in Metabolism & Endocrine research.